Stampa…

Poly(methyl vinyl ether) 30% in water

Fornitore: TCI EUROPE

P0384-25G P0384-500G
TCIAP0384-25GEA 24 EUR
TCIAP0384-25G TCIAP0384-500G
Poly(methyl vinyl ether) 30% in water
Poly(methyl vinyl ether)
Formula: C₃H₆O
MDL Number: MFCD00161528
CAS Number: 9003-09-2

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown clear liquid to slightly cloudy liquid
Evaporation residue 27.0 to 33.0 %
Viscosity 2000 to 4000 mPa-s
Absorbance max. 0.300 (400.0 nm)
Absorbance2 max. 0.050 (600.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR