Stampa…

Poly(1-vinylpyrrolidone-co-vinyl acetate) (copolymer 7:3) 50% in ethanol

Fornitore: TCI EUROPE
Pericolo

P0382-500G P0382-25G
TCIAP0382-500GEA 70 EUR
TCIAP0382-500G TCIAP0382-25G
Poly(1-vinylpyrrolidone-co-vinyl acetate) (copolymer 7:3) 50% in ethanol
Poly(1-vinylpyrrolidone-co-vinyl acetate)
MDL Number: MFCD00134018
CAS Number: 25086-89-9
UN: 1170
ADR: 3,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Total Nitrogen 4.2 to 4.8 %
Evaporation residue 46.0 to 54.0 %
Viscosity 800.0 to 3,000.0 mPa-s

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR