Stampa…

Phenylthiohydantoin-tyrosine ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 5-(4-Hydroxybenzyl)-3-phenyl-2-thioxoimidazolidin-4-one, PTH-tyrosine, 5-(4-Hydroxybenzyl)-3-phenyl-2-thiohydantoin, Phenylthiohydantoin-tyrosine

P0368-1G P0368-100MG
TCIAP0368-1GEA 127 EUR
TCIAP0368-1G TCIAP0368-100MG
Phenylthiohydantoin-tyrosine ≥98.0% (by HPLC)
Phenylthiohydantoin-tyrosine
Formula: C₁₆H₁₄N₂O₂S
Peso molecolare: 298,36 g/mol
MDL Number: MFCD00027415
CAS Number: 4332-95-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystaline
Purity (HPLC) min. 98.0 area%
Melting point 217.0 to 225.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR