Stampa…

Phenylthiohydantoin-phenylalanine

Fornitore: TCI

Synonyms: 5-Benzyl-3-phenyl-2-thiohydantoin, PTH-phenylalanine

P0367-1G P0367-100MG
TCIAP0367-1GEA 82 EUR
TCIAP0367-1G TCIAP0367-100MG
Phenylthiohydantoin-phenylalanine
Phenylthiohydantoin-phenylalanine
Formula: C₁₆H₁₄N₂OS
Peso molecolare: 282.36 g/mol
MDL Number: MFCD01076623
CAS Number: 4332-97-2
EINECS: 224-372-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 187.0 to 191.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR