Phenylthiohydantoinaspartic acid ≥95.0% (by titrimetric analysis)
Fornitore: TCI
Synonyms:
PTH-aspartic Acid, 3-Phenyl-2-thiohydantoin-5-acetic Acid
Formula:
C₁₁H₁₀N₂O₃S Peso molecolare: 250.27 g/mol |
MDL Number:
MFCD01076622 CAS Number: 5624-13-5 EINECS: 227-055-5 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...