Stampa…

2-(2-Pyridyl)ethanesulphonic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

P0352-25G P0352-5G
TCIAP0352-25GEA 98 EUR
TCIAP0352-25G TCIAP0352-5G
2-(2-Pyridyl)ethanesulphonic acid ≥98.0% (by HPLC, titration analysis)
2-(2-Pyridyl)ethanesulphonic acid
Formula: C₇H₉NO₃S
MDL Number: MFCD00006365
CAS Number: 68922-18-9
EINECS: 272-989-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6c59eff02169d9b94d542c34b60943e438b4e3e&vid=f6c59eff02169d9b94d542c34b60943e438b4e3e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR