4-Pyridinepropanol ≥97.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
P0351-25G
P0351-5G
TCIAP0351-25GEA
364
EUR
TCIAP0351-25G
TCIAP0351-5G
4-Pyridinepropanol ≥97.0% (by GC, titration analysis)
4-Pyridinepropanol
Formula:
C₈H₁₁NO Punto di ebollizione: 289°C |
MDL Number:
MFCD00047457 CAS Number: 2629-72-3 EINECS: 220-107-8 |
Risultati del test di specificazione
Appearance | White to Gray to Brown powder to lump |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...