Stampa…

1-Phenyl-2-pyrrolidone ≥99.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: 1-Phenyl-2-pyrrolidinone

P0319-25G P0319-5G
TCIAP0319-25GEA 77 EUR
TCIAP0319-25G TCIAP0319-5G
1-Phenyl-2-pyrrolidone ≥99.0% (by GC)
1-Phenyl-2-pyrrolidone
Formula: C₁₀H₁₁NO
MDL Number: MFCD00003192
CAS Number: 4641-57-0
EINECS: 225-069-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a912fca9e3af43cbd72dd9da8ef457a628650a95&vid=a912fca9e3af43cbd72dd9da8ef457a628650a95&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 66.0 to 70.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR