Ditridecyl phthalate (mixture of branched chain isomers)
Fornitore: TCI EUROPE
Synonyms:
Phthalic Acid Ditridecyl Ester
P0307-25G
P0307-500G
TCIAP0307-25GEA
17
EUR
TCIAP0307-25G
TCIAP0307-500G
Ditridecyl phthalate (mixture of branched chain isomers)
Ditridecyl phthalate (mixture of branched chain isomers)
Formula:
C₃₄H₅₈O₄ Peso molecolare: 530,83 g/mol Flash Pt: 242°C |
MDL Number:
MFCD00048451 CAS Number: 75359-31-8 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | multi component mixture |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...