Stampa…

Diisodecyl phthalate (mixture of branched chain isomers)

Fornitore: TCI EUROPE
Attenzione

Synonyms: DIDP, Phthalic acid diisodecyl ester

P0299-25ML P0299-500ML
TCIAP0299-25MLEA 16 EUR
TCIAP0299-25ML TCIAP0299-500ML
Diisodecyl phthalate (mixture of branched chain isomers)
Diisodecyl phthalate
Formula: C₂₈H₄₆O₄
Peso molecolare: 446,67 g/mol
Flash Pt: 235°C
MDL Number: MFCD00048389
CAS Number: 26761-40-0
EINECS: 247-977-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=070004447c8dbd4817bd780ff3dd97269e1d06ad&vid=070004447c8dbd4817bd780ff3dd97269e1d06ad&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Ester Value 246 to 254

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR