Dibutyl phosphate ≥97.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Dibutylphosphate, Phosphoric Acid Dibutyl Ester
P0260-25G
P0260-500G
TCIAP0260-25GEA
19
EUR
TCIAP0260-25G
TCIAP0260-500G
Dibutyl phosphate ≥97.0% (by titrimetric analysis)
Dibutyl phosphate
Formula:
C₈H₁₉O₄P Peso molecolare: 210,21 g/mol Punto di ebollizione: 138°C Punto di fusione: –13°C Flash Pt: 196°C |
MDL Number:
MFCD00015245 CAS Number: 107-66-4 EINECS: 203-509-8 UN: 1718 ADR: 8,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...