Stampa…

Phenyl-p-toluenesulphonate ≥99.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: p-Toluenesulphonic acid Phenyl Ester

P0239-10G P0239-25G
TCIAP0239-10GEA 66 EUR
TCIAP0239-10G TCIAP0239-25G
Phenyl-p-toluenesulphonate ≥99.0% (by GC)
Phenyl-p-toluenesulphonate
Formula: C₁₃H₁₂O₃S
Peso molecolare: 248,30 g/mol
MDL Number: MFCD00025996
CAS Number: 640-60-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 94.0 to 97.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR