Stampa…

3-Phenylsalicylic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Hydroxy-[1,1'-biphenyl]-3-carboxylic acid, 2-Hydroxybiphenyl-3-carboxylic Acid

P0226-25G P0226-5G
TCIAP0226-25GEA 188 EUR
TCIAP0226-25G TCIAP0226-5G
3-Phenylsalicylic acid ≥98.0% (by titrimetric analysis)
3-Phenylsalicylic acid
Formula: C₁₃H₁₀O₃
Peso molecolare: 214,22 g/mol
MDL Number: MFCD00020257
CAS Number: 304-06-3
EINECS: 206-148-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cee5b04b7d6d514b91e349e52f2ef628f59ef41f&vid=cee5b04b7d6d514b91e349e52f2ef628f59ef41f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 189.0 to 193.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR