Methyl phenoxyacetate ≥99.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Phenoxyacetic Acid Methyl Ester, Methyl 2-phenoxyacetate
P0110-25G
P0110-500G
TCIAP0110-25GEA
92
EUR
TCIAP0110-25G
TCIAP0110-500G
Methyl phenoxyacetate ≥99.0% (by GC)
Methyl phenoxyacetate
Formula:
C₉H₁₀O₃ Peso molecolare: 166,18 g/mol Punto di ebollizione: 245°C Flash Pt: 111°C |
MDL Number:
MFCD00010227 CAS Number: 2065-23-8 EINECS: 218-176-4 |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...