Stampa…

Sodium-4-hydroxybenzenesulphonate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Sodium p-Phenolsulphonate, 4-Hydroxybenzenesulphonic acid Sodium Salt, p-Phenolsulphonic acid Sodium Salt

P0105-25G P0105-500G
TCIAP0105-25GEA 18 EUR
TCIAP0105-25G TCIAP0105-500G
Sodium-4-hydroxybenzenesulphonate ≥98.0% (by HPLC, titration analysis)
Sodium-4-hydroxybenzenesulphonate
Formula: C₆H₆O₄S
Peso molecolare: 196,15 g/mol
MDL Number: MFCD00044734
CAS Number: 825-90-1
EINECS: 212-550-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 % (calcd.on anh.substance)
Water max. 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR