Stampa…

4-Hydroxybenzenesulphonic acid ≥85.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

P0104-25G P0104-500G
TCIAP0104-25GEA 16 EUR
TCIAP0104-25G TCIAP0104-500G
4-Hydroxybenzenesulphonic acid ≥85.0% (by titrimetric analysis)
4-Hydroxybenzenesulphonic acid
Formula: C₆H₆O₄S
MDL Number: MFCD00673723
CAS Number: 98-67-9
EINECS: 202-691-6
UN: 1759
ADR: 8,III
Merck Index: 14,07246

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to lump
Purity (Neutralization titration) min. 85.0% (calcd.on anh.substance)
Water max. 15.0 %
Sulfate max. 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR