Stampa…

Sodium-D-pantothenate ≥97.0% (by total nitrogen basis)

Fornitore: TCI EUROPE

Synonyms: D-Pantothenic Acid Sodium Salt

P0013-25G P0013-250G
TCIAP0013-25GEA 81 EUR
TCIAP0013-25G TCIAP0013-250G
Sodium-D-pantothenate ≥97.0% (by total nitrogen basis)
Sodium-D-pantothenate
Formula: C₉H₁₇NO₅
Peso molecolare: 241,22 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00002767
CAS Number: 867-81-2
EINECS: 212-768-6

Visualizza i codici

Risultati del test di specificazione

Appearance White powder to crystal
Purity (with Total Nitrogen) min. 97.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +25.0 to +28.5 deg (C=5, H2O) (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 5.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR