Oxetan-3-ylmethanol ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
3-(Hydroxymethyl)oxetane, 3-Oxetanemethanol, Oxetan-3-ylmethanol
O0558-1G
O0558-200MG
TCIAO0558-1GEA
85
EUR
TCIAO0558-1G
TCIAO0558-200MG
Oxetan-3-ylmethanol ≥98.0% (by GC)
Oxetan-3-ylmethanol
Formula:
C₄H₈O₂ Peso molecolare: 88,11 g/mol Flash Pt: 106°C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD17015894 CAS Number: 6246-06-6 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...