Sodium 5-(Trifluoromethyl)pyridine-2-sulphinate ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
5-(Trifluoromethyl)pyridine-2-sulphinic acid sodium salt
O0546-250MG
O0546-1G
TCIAO0546-250MGEA
69
EUR
TCIAO0546-250MG
TCIAO0546-1G
Sodium 5-(Trifluoromethyl)pyridine-2-sulphinate ≥98.0% (by HPLC, titration analysis)
Sodium 5-(Trifluoromethyl)pyridine-2-sulphinate
Formula:
C₆H₃F₃NNaO₂S Peso molecolare: 233,14 g/mol |
MDL Number:
MFCD31442692 CAS Number: 2098851-48-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...