Stampa…

Sodium Pyrimidine-2-sulphinate ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Sodium pyrimidine-2-sulphinate

O0541-1G O0541-250MG
TCIAO0541-1GEA 295 EUR
TCIAO0541-1G TCIAO0541-250MG
Sodium Pyrimidine-2-sulphinate ≥97.0% (by HPLC, titration analysis)
Sodium Pyrimidine-2-sulphinate
Formula: C₄H₃N₂NaO₂S
Peso molecolare: 166,13 g/mol
MDL Number: MFCD32068077
CAS Number: 2188151-68-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR