5-Oxotetrahydrofuran-3-yl methacrylate ≥97.0% (by GC) stabilizzato
Fornitore: TCI EUROPE
Synonyms:
beta-Hydroxy-gamma-butyrolactone methacrylate, Methacrylic acid 5-Oxotetrahydrofuran-3-yl ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
O0538-5G
O0538-25G
TCIAO0538-5GEA
57
EUR
TCIAO0538-5G
TCIAO0538-25G
5-Oxotetrahydrofuran-3-yl methacrylate ≥97.0% (by GC) stabilizzato
5-Oxotetrahydrofuran-3-yl methacrylate
Formula:
C₈H₁₀O₄ Peso molecolare: 170,16 g/mol Temperatura di stoccaggio: Refrigerata |
CAS Number:
130224-95-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...