Stampa…

Olmesartan medoxomil ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl 1-((2'-(2H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-4-(2-hydroxypropan-2-yl)-2-propyl-1H-imidazole-5-carboxylate, (5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl-1-((2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)-4-(2-hydroxypropan-2-yl)-2-propyl-1H-imidazole-5-carboxylate

O0510-1G O0510-5G
TCIAO0510-1GEA 59 EUR
TCIAO0510-1G TCIAO0510-5G
Olmesartan medoxomil ≥98.0% (by HPLC, titration analysis)
Olmesartan medoxomil
Formula: C₂₉H₃₀N₆O₆
Peso molecolare: 558,60 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00944911
CAS Number: 144689-63-4
Merck Index: 14,06839

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR