Stampa…

Oxibendazole ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI EUROPE

Synonyms: (5-Propoxy-1H-benzimidazol-2-yl)carbamic acid methyl ester, Methyl (5-Propoxy-1H-benzo[d]imidazol-2-yl)carbamate

O0499-100MG O0499-500MG
TCIAO0499-100MGEA 92 EUR
TCIAO0499-100MG TCIAO0499-500MG
Oxibendazole ≥98.0% (by HPLC, total nitrogen)
Oxibendazole
Formula: C₁₂H₁₅N₃O₃
Peso molecolare: 249,27 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00133728
CAS Number: 20559-55-1
EINECS: 243-877-7
Merck Index: 14,06936

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR