Sodio tetrafluoroborato ≥95.0% (by titrimetric analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
BF₄Na Punto di fusione: 384°C |
MDL Number:
MFCD00003515 CAS Number: 13755-29-8 EINECS: 237-340-6 Merck Index: 14,08617 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...