Stampa…

Octavinyloctasilasesquioxane

Fornitore: TCI EUROPE

Synonyms: 1,3,5,7,9,11,13,15-Octavinylpentacyclo[9.5.1.1(3,9).1(5,15).1(7,13)]octasiloxane, 1,3,5,7,9,11,13,15-Octavinyl-2,4,6,8,10,12,14,16,17,18,19,20-dodecaoxa-1,3,5,7,9,11,13,15-octasilapentacyclo[9.5.1.13,9.15,15.17,13]icosane

O0491-1G O0491-5G
TCIAO0491-1GEA 59 EUR
TCIAO0491-1G TCIAO0491-5G
Octavinyloctasilasesquioxane
Octavinyloctasilasesquioxane
Formula: C₁₆H₂₄O₁₂Si₈
Peso molecolare: 633,04 g/mol
MDL Number: MFCD24450214
CAS Number: 69655-76-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
IR to pass test
NMR to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR