Stampa…

4,5,7,8,12,13,15,16-Octafluoro[2.2]paracyclophane ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Parylene F Dimer, 5,6,11,12,13,14,15,16-Octafluorotricyclo[8.2.2.2(4,7)]hexadeca-4,6,10,12,13,15-hexaene

O0455-1G O0455-5G
TCIAO0455-1GEA 42 EUR
TCIAO0455-1G TCIAO0455-5G
4,5,7,8,12,13,15,16-Octafluoro[2.2]paracyclophane ≥98.0% (by GC)
4,5,7,8,12,13,15,16-Octafluoro[2.2]paracyclophane
Formula: C₁₆H₈F₈
Peso molecolare: 352,23 g/mol
MDL Number: MFCD13195584
CAS Number: 1785-64-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7964bfbae46520c0d7c65eb4c451cf846314728&vid=b7964bfbae46520c0d7c65eb4c451cf846314728&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR