Stampa…

1-Octylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI
Attenzione

O0452-1G O0452-5G
TCIAO0452-1GEA 28 EUR
TCIAO0452-1G TCIAO0452-5G
1-Octylboronic acid (contains varying amounts of Anhydride)
1-Octylboronic acid
Formula: C₈H₁₉BO₂
Punto di fusione: 81°C
MDL Number: MFCD01074560
CAS Number: 28741-08-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=127fa7a37457383496940ddaf11adbb219e6bd4d&vid=127fa7a37457383496940ddaf11adbb219e6bd4d&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR