3-Oxabicyclo[3.1.0]hexane-2,4-dione ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Cyclopropane-1,2-dicarboxylic acid anhydride
O0434-5G
O0434-1G
TCIAO0434-5GEA
184
EUR
TCIAO0434-5G
TCIAO0434-1G
3-Oxabicyclo[3.1.0]hexane-2,4-dione ≥98.0% (by GC, titration analysis)
3-Oxabicyclo[3.1.0]hexane-2,4-dione
Formula:
C5H4O3 Peso molecolare: 112,08 g/mol Punto di ebollizione: 114°C |
MDL Number:
MFCD00126929 CAS Number: 5617-74-3 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Methanolysis Method) | min. 98.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...