Ozagrel hydrochloride hydrate ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
Ozagrel HCl, (E)-3-(4-((1H-Imidazol-1-yl)methyl)phenyl)acrylic acid hydrochloride, trans-4-[(1-Imidazolyl)methyl]cinnamic Acid Hydrochloride, (E)-3-[4-(Imidazol-1-ylmethyl)phenyl]propenoic Acid Hydrochloride
O0419-1G
O0419-5G
TCIAO0419-1GEA
97
EUR
TCIAO0419-1G
TCIAO0419-5G
Ozagrel hydrochloride hydrate ≥98.0% (by HPLC, titration analysis)
Ozagrel hydrochloride hydrate
Formula:
C₁₃H₁₂N₂O₂ Peso molecolare: 264,71 g/mol Punto di fusione: 217°C |
MDL Number:
MFCD06795644 CAS Number: 78712-43-3 Merck Index: 14,06980 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % (calcd.on anh.substance) |
Water | 5.0 to 8.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...