4-n-Octyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥96.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
4-n-Octyl-2-thiopheneboronic Acid Pinacol Ester, 4,4,5,5-Tetramethyl-2-(4-n-octyl-2-thienyl)-1,3,2-dioxaborolan
O0411-1G
O0411-5G
TCIAO0411-1GEA
101
EUR
TCIAO0411-1G
TCIAO0411-5G
4-n-Octyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene ≥96.0% (by GC)
4-n-Octyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene
Formula:
C₁₈H₃₁BO₂S Peso molecolare: 322,31 g/mol Temperatura di stoccaggio: Refrigerata |
CAS Number:
405165-12-0 |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...