Stampa…

Ondansetron hydrochloride dihydrate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 9-Methyl-3-((2-methyl-1H-imidazol-1-yl)methyl)-1,2,3,9-tetrahydro-4H-carbazol-4-one hydrochloride dihydrate, GR 3832 HCl 2H2O, 1,2,3,9-Tetrahydro-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-4H-carbazol-4-one Hydrochloride, SN-37 HCl 2H2O, NSC665799 HCl 2H2O

O0407-1G O0407-5G
TCIAO0407-1GEA 52 EUR
TCIAO0407-1G TCIAO0407-5G
Ondansetron hydrochloride dihydrate ≥98.0% (by HPLC, titration analysis)
Ondansetron hydrochloride dihydrate
Formula: C₁₈H₁₉N₃O·HCl·₂H₂O
Peso molecolare: 365,86 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00374371
CAS Number: 103639-04-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=303b043daeeffda65d011456d5c62998ad5c7913&vid=303b043daeeffda65d011456d5c62998ad5c7913&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 % (calcd.on anh.substance)
Melting point 176.0 to 180.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR