Stampa…

Ofloxacin ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 9-Fluoro-3-methyl-10-(4-methylpiperazin-1-yl)-7-oxo-3,7-dihydro-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid, 9-Fluoro-3,7-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-2H-[1,4]oxazino[2,3,4-ij]quinoline-6-carboxylic acid

O0403-5G O0403-25G
TCIAO0403-5GEA 52 EUR
TCIAO0403-5G TCIAO0403-25G
Ofloxacin ≥98.0% (by HPLC, titration analysis)
Ofloxacin
Formula: C₁₈H₂₀FN₃O₄
Peso molecolare: 361,37 g/mol
MDL Number: MFCD00226105
CAS Number: 82419-36-1
Merck Index: 14,06771
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b678f39cebf92fd14a5eff55fa5078405804de9&vid=5b678f39cebf92fd14a5eff55fa5078405804de9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR