Stampa…

Oxaprozin ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 3-(4,5-Diphenyloxazol-2-yl)propanoic acid, 4,5-Diphenyl-2-oxazolepropionic Acid

O0377-5G O0377-25G
TCIAO0377-5GEA 44 EUR
TCIAO0377-5G TCIAO0377-25G
Oxaprozin ≥98.0% (by HPLC, titration analysis)
Oxaprozin
Formula: C₁₈H₁₅NO₃
Peso molecolare: 293,32 g/mol
MDL Number: MFCD00215977
CAS Number: 21256-18-8
EINECS: 244-296-1
Merck Index: 14,06924

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 161.0 to 165.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR