Oxyresveratrol ≥96.0% (by GC)
Fornitore: TCI
|
Attenzione
|
O0373-1G
O0373-100MG
TCIAO0373-1GEA
373
EUR
TCIAO0373-1G
TCIAO0373-100MG
Oxyresveratrol ≥96.0% (by GC)
Oxyresveratrol
Formula:
C₁₄H₁₂O₄ |
MDL Number:
MFCD11974969 CAS Number: 29700-22-9 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...