Oxacillin sodium salt monohydrate ≥98.0% (by HPLC)
Fornitore: TCI
O0353-25G
O0353-5G
O0353-1G
TCIAO0353-25GEA
203
EUR
TCIAO0353-25G
TCIAO0353-5G
TCIAO0353-1G
Oxacillin sodium salt monohydrate ≥98.0% (by HPLC)
Oxacillin sodium salt monohydrate
Formula:
C₁₉H₁₉N₃O₅S Peso molecolare: 423.42 g/mol Punto di fusione: 188°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00167146 CAS Number: 7240-38-2 Merck Index: 14,06904 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Specific rotation [a]20/D | +200.0 to +210.0 deg (C=1, H2O) (calcd.on anh.substance) |
Water | 3.5 to 5.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...