Stampa…

Octaphenyloctasilsesquioxane

Fornitore: TCI EUROPE

Synonyms: 1,3,5,7,9,11,13,15-Octaphenyl-2,4,6,8,10,12,14,16,17,18,19,20-dodecaoxa-1,3,5,7,9,11,13,15-octasilapentacyclo[9.5.1.13,9.15,15.17,13]icosane, 1,3,5,7,9,11,13,15-Octaphenylpentacyclo[9.5.1.1(3,9).1(5,15).1(7,13)]octasiloxane

O0349-5G O0349-25G
TCIAO0349-5GEA 113 EUR
TCIAO0349-5G TCIAO0349-25G
Octaphenyloctasilsesquioxane
Octaphenyloctasilsesquioxane
Formula: C₄₈H₄₀O₁₂Si₈
Peso molecolare: 1033,52 g/mol
Punto di fusione: 500°C
MDL Number: MFCD00308870
CAS Number: 5256-79-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af7509f72d1df1b1f1470ddd9cf4599b26c03de5&vid=af7509f72d1df1b1f1470ddd9cf4599b26c03de5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
IR to pass test
NMR to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR