Bis[3,4,6-trichloro-2-(pentyloxycarbonyl)phenyl]oxalate ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Bis[2,4,5-trichloro-6-(pentyloxycarbonyl)phenyl] Oxalate, Bis[3,4,6-trichloro-2-(pentyloxycarbonyl)phenyl] Oxalate, Oxalic Acid Bis[2,4,5-trichloro-6-(pentyloxycarbonyl)phenyl] Ester, Oxalic Acid Bis[3,4,6-trichloro-2-(pentyloxycarbonyl)phenyl] Ester
Formula:
C₂₆H₂₄Cl₆O₈ Peso molecolare: 677,17 g/mol |
MDL Number:
MFCD00191674 CAS Number: 30431-54-0 EINECS: 250-195-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 81.0 to 84.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...