Stampa…

Diethyl-3-oxopimelate ≥96.0% (by titrimetric analysis)

Fornitore: TCI

Synonyms: 3-Oxopimelic Acid Diethyl Ester, 3-Oxoheptanedioic Acid Diethyl Ester, Diethyl 3-Oxoheptanedioate

O0229-1G O0229-5G
TCIAO0229-1GEA 44 EUR
TCIAO0229-1G TCIAO0229-5G
Diethyl-3-oxopimelate ≥96.0% (by titrimetric analysis)
Diethyl-3-oxopimelate
Formula: C₁₁H₁₈O₅
Peso molecolare: 230.26 g/mol
MDL Number: MFCD00009214
CAS Number: 40420-22-2
EINECS: 254-912-0

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR