Methyl trans-9-octadecenoate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Elaidic Acid Methyl Ester, trans-9-Octadecenoic Acid Methyl Ester, (E)-Methyl octadec-9-enoate, Methyl Elaidate
O0226-5ML
O0226-1ML
TCIAO0226-5MLEA
122
EUR
TCIAO0226-5ML
TCIAO0226-1ML
Methyl trans-9-octadecenoate ≥98.0% (by GC)
Methyl trans-9-octadecenoate
Formula:
C₁₉H₃₆O₂ Peso molecolare: 296,50 g/mol Punto di ebollizione: 220°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00066521 CAS Number: 1937-62-8 EINECS: 217-712-4 |
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...