n-Octyl phenyl ether ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
n-Octyloxybenzene
O0224-10ML
O0224-25ML
TCIAO0224-10MLEA
104
EUR
TCIAO0224-10ML
TCIAO0224-25ML
n-Octyl phenyl ether ≥98.0% (by GC)
n-Octyl phenyl ether
Formula:
C₁₄H₂₂O Peso molecolare: 206,33 g/mol Punto di ebollizione: 134°C |
MDL Number:
MFCD00191564 CAS Number: 1818-07-1 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...