Sodium-4-n-octylbenzenesulphonate ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₄H₂₂O₃S |
MDL Number:
MFCD00007512 CAS Number: 6149-03-7 EINECS: 228-158-8 |
Risultati del test di specificazione
Appearance | White powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Drying loss | max. 2.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...