Stampa…

Oleamide >65.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Oleyramide, cis-9-Octadecenamide

O0107-25G O0107-500G
TCIAO0107-25GEA 17 EUR
TCIAO0107-25G TCIAO0107-500G
Oleamide >65.0% (by GC)
Oleamide
Formula: C₁₈H₃₅NO
Peso molecolare: 281,48 g/mol
MDL Number: MFCD00053638
CAS Number: 301-02-0
EINECS: 206-103-9

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 65.0 %
Melting point 65.0 to 75.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR