Dioctyl ether ≥96.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
n-Octyl ether, di-n-Octyl ether, Dioctylether
O0047-25ML
O0047-500ML
O0047-100ML
TCIAO0047-25MLEA
30
EUR
TCIAO0047-25ML
TCIAO0047-500ML
TCIAO0047-100ML
Dioctyl ether ≥96.0% (by GC)
Dioctyl ether
Formula:
C₁₆H₃₄O Peso molecolare: 242,45 g/mol Punto di ebollizione: 287°C Punto di fusione: -7°C |
MDL Number:
MFCD00009563 CAS Number: 629-82-3 EINECS: 211-112-6 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...