Stampa…

n-Octanoic anhydride ≥95.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

Synonyms: n-Octanoic Anhydride, Caprylic anhydride, Octanoic anhydride

O0035-25ML O0035-500ML
TCIAO0035-25MLEA 42 EUR
TCIAO0035-25ML TCIAO0035-500ML
n-Octanoic anhydride ≥95.0% (by GC, titration analysis)
n-Octanoic anhydride
Formula: C₁₆H₃₀O₃
Punto di ebollizione: 180°C
Punto di fusione: -1°C
MDL Number: MFCD00056161
CAS Number: 623-66-5
EINECS: 210-806-6
UN: 3265
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Morpholine Method) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR