Amyl-n-octanoate ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Pentyl n-Octanoate, Pentyl octanoate, n-Octanoic Acid Amyl Ester
O0028-25ML
O0028-500ML
TCIAO0028-25MLEA
16
EUR
TCIAO0028-25ML
TCIAO0028-500ML
Amyl-n-octanoate ≥98.0% (by GC)
Amyl-n-octanoate
Formula:
C₁₃H₂₆O₂ Peso molecolare: 214,35 g/mol Punto di ebollizione: 260°C |
MDL Number:
MFCD00048919 CAS Number: 638-25-5 EINECS: 211-328-0 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...