Stampa…

Octadecyl vinyl ether ≥85.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Stearyl Vinyl Ether

O0019-25ML O0019-500ML
TCIAO0019-25MLEA 16 EUR
TCIAO0019-25ML TCIAO0019-500ML
Octadecyl vinyl ether ≥85.0% (by GC)
Octadecyl vinyl ether
Formula: C₂₀H₄₀O
Peso molecolare: 296,54 g/mol
MDL Number: MFCD00026686
CAS Number: 930-02-9
EINECS: 213-208-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to lump
Purity (GC) min. 85.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR