Stampa…

Naratriptan hydrochloride ≥98% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N-Methyl-3-(1-methyl-4-piperidinyl)-1H-indole-5-ethanesulphonamide hydrochloride

N1153-250MG N1153-50MG
TCIAN1153-250MGEA 403 EUR
TCIAN1153-250MG TCIAN1153-50MG
Naratriptan hydrochloride ≥98% (by GC, titration analysis)
Naratriptan hydrochloride
Formula: C₁₇H₂₅N₃O₂S·HCl
Peso molecolare: 371,92 g/mol
MDL Number: MFCD08141821
CAS Number: 143388-64-1
Merck Index: 14,06420

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 245.0 to 249.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR