Stampa…

4-(1-Naphthyl)-3,5-diphenyl-1,2,4-triazole ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(1-Naphthyl)-3,5-diphenyl-1,2,4-triazol

N1139-1G N1139-200MG
TCIAN1139-1GEA 219 EUR
TCIAN1139-1G TCIAN1139-200MG
4-(1-Naphthyl)-3,5-diphenyl-1,2,4-triazole ≥98.0% (by GC)
4-(1-Naphthyl)-3,5-diphenyl-1,2,4-triazole
Formula: C₂₄H₁₇N₃
Peso molecolare: 347,42 g/mol
MDL Number: MFCD09832971
CAS Number: 16152-10-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 243.0 to 247.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR