Nocodazole ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl (6-(thiophene-2-carbonyl)-1H-benzo[d]imidazol-2-yl)carbamate
N1138-25MG
N1138-100MG
TCIAN1138-25MGEA
150
EUR
TCIAN1138-25MG
TCIAN1138-100MG
Nocodazole ≥98.0% (by HPLC)
Nocodazole
Formula:
C₁₄H₁₁N₃O₃S Peso molecolare: 301,32 g/mol Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD00005588 CAS Number: 31430-18-9 EINECS: 250-626-5 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...