4-(Naphthalen-2-yl)-N-phenylaniline ≥98.0% (by GC)
Fornitore: TCI
N1134-1G
N1134-5G
TCIAN1134-1GEA
68
EUR
TCIAN1134-1G
TCIAN1134-5G
4-(Naphthalen-2-yl)-N-phenylaniline ≥98.0% (by GC)
4-(Naphthalen-2-yl)-N-phenylaniline
Formula:
C₂₂H₁₇N |
MDL Number:
MFCD30478546 CAS Number: 897671-79-3 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 152.0 to 156.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...